siRNA Details
Gene Symbol: Cypt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039941.2 | NP_001035030.1 | 1 | 289 | ||
GenBank mRNA | AM040457.1 | - | 1 | 289 |
Gene Symbol: Cypt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016467.1 | - | 1 | 297 | ||
AM040451.1 | - | 1 | 289 |
