siRNA Details
Gene Symbol: Cypt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039944.2 | NP_001035033.1 | 1 | 197 | ||
GenBank mRNA | AM040459.1 | - | 1 | 197 |
Gene Symbol: Cypt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173412.2 | NP_775588.2 | 1 | 210 | ||
GenBank mRNA | AF463501.1 | - | 1 | 210 | ||
AM040453.1 | - | 1 | 194 | |||
BC048530.1 | - | 1 | 199 | |||
BC060952.1 | - | 1 | 199 | 6772242 | ||
BC147783.1 | - | 1 | 294 | 9087502 |
Gene Symbol: Cypt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039942.2 | NP_001035031.1 | 1 | 194 | ||
GenBank mRNA | AM040458.1 | - | 1 | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
