siRNA Details
Gene Symbol: Tmem238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029384.1 | NP_083660.1 | 2 | 1005 | ||
GenBank mRNA | AK008900.1 | - | 2 | 1009 | ||
BC132101.1 | - | 2 | 1101 | 40130378 | ||
BC144965.1 | - | 2 | 1101 | 9053513 |
