siRNA Details
Gene Symbol: Isoc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101598.1 | NP_001095068.1 | 2 | 228 | ||
GenBank mRNA | BC158139.1 | - | 2 | 128 | 9007402 | |
BG404719.1 | - | 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
