siRNA Details
Gene Symbol: Sms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009214.3 | NP_033240.3 | 8 | 860 | ||
GenBank mRNA | AF031486.1 | - | 8 | 843 | ||
AK017775.1 | - | 8 | 831 | |||
AK049787.1 | - | 8 | 849 | |||
AK136798.1 | - | 8 | 861 | |||
AK149929.1 | - | 8 | 858 | |||
AK160659.1 | - | 8 | 861 | |||
AK166665.1 | - | 8 | 886 | |||
BC046623.1 | - | 8 | 842 | 6315420 | ||
BC058688.1 | - | 8 | 1101 | 6413869 | ||
Y09419.1 | - | 8 | 833 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
