siRNA Details
Gene Symbol: Rhox3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102457.4 | NP_001095927.3 | 4 | 765 | ||
GenBank mRNA | BC087875.1 | - | 3 | 590 | ||
BC092527.1 | - | 3 | 409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
