siRNA Details
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008997.3 | NP_033023.1 | 5 | 1829 | ||
GenBank mRNA | AK146452.1 | - | 5 | 1690 | ||
AK146460.1 | - | 5 | 1641 | |||
AK146623.1 | - | 5 | 1641 | |||
AK146978.1 | - | 5 | 1621 | |||
AK147077.1 | - | 5 | 1660 | |||
AK148022.1 | - | 5 | 1703 | |||
BC054753.1 | - | 5 | 1645 | 6411651 | ||
BC085270.1 | - | 5 | 1639 | 6478784 | ||
L26528.1 | - | 5 | 1641 |
