siRNA Details
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008831.4 | NP_032857.1 | 7 | 1002 | ||
GenBank mRNA | AK010619.1 | - | 7 | 945 | ||
AK150956.1 | - | 7 | 947 | |||
AK151073.1 | - | 7 | 947 | |||
AK152933.1 | - | 7 | 948 | |||
AK160490.1 | - | 7 | 644 | |||
AK168248.1 | - | 7 | 950 | |||
AK168656.1 | - | 7 | 941 | |||
AK180942.1 | - | 1 | 63 | |||
AK190335.1 | - | 1 | 63 | |||
BC083354.1 | - | 6 | 924 | 5715690 | ||
BC089034.1 | - | 6 | 926 | 30675988 |
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161411.1 | - | 1 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
