siRNA Details
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008831.4 | NP_032857.1 | 4 | 431 | ||
GenBank mRNA | AK002714.1 | - | 3 | 377 | ||
AK010619.1 | - | 4 | 374 | |||
AK150956.1 | - | 4 | 376 | |||
AK151073.1 | - | 4 | 376 | |||
AK152933.1 | - | 4 | 378 | |||
AK160490.1 | - | 4 | 370 | |||
AK168248.1 | - | 4 | 379 | |||
AK168656.1 | - | 4 | 370 | |||
AK190324.1 | - | 1 | 58 | |||
BC083354.1 | - | 3 | 353 | 5715690 | ||
BC089034.1 | - | 3 | 355 | 30675988 | ||
X78682.1 | - | 3 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
