siRNA Details
Gene Symbol: 2810047C21Rik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012911.1 | - | 342 | |||
AK028951.1 | - | 652 | ||||
AK045648.1 | - | 643 | ||||
AK083498.1 | - | 6 | 838 | |||
BC071238.1 | - | 611 |

More Information
Other Species Targeted

