siRNA Details
Gene Symbol: Pcnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024622.2 | NP_001019793.1 | 3 | 357 | ||
GenBank mRNA | AK145195.1 | - | 3 | 316 | ||
AK150595.1 | - | 3 | 318 | |||
AK152279.1 | - | 3 | 320 | |||
AK159201.1 | - | 3 | 328 | |||
AK166071.1 | - | 3 | 341 | |||
AK168723.1 | - | 3 | 285 | |||
AK172325.1 | - | 2 | 362 | |||
BC023320.1 | - | 3 | 328 | |||
BC061235.1 | - | 3 | 321 | 6593866 | ||
BC106192.1 | - | 3 | 266 | |||
BC132588.1 | - | 3 | 379 | 40130865 | ||
BC132590.1 | - | 3 | 379 | 40130867 | ||
BC145194.1 | - | 3 | 327 | |||
BY287179.1 | - | 3 | 357 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001047846, XM_006248241, XM_006248242, XM_008768631, XM_008768630, and others.
Gene Ontology Categories:
