siRNA Details
Gene Symbol: Pcnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024622.2 | NP_001019793.1 | 5 | 540 | ||
GenBank mRNA | AK145195.1 | - | 5 | 499 | ||
AK150595.1 | - | 5 | 501 | |||
AK152279.1 | - | 5 | 503 | |||
AK159201.1 | - | 5 | 511 | |||
AK168723.1 | - | 5 | 468 | |||
AK172325.1 | - | 4 | 545 | |||
BC023320.1 | - | 5 | 511 | |||
BC061235.1 | - | 5 | 504 | 6593866 | ||
BC106192.1 | - | 5 | 449 | |||
BC132588.1 | - | 5 | 562 | 40130865 | ||
BC132590.1 | - | 5 | 562 | 40130867 | ||
BC145194.1 | - | 5 | 510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
