siRNA Details
Gene Symbol: Vmn2r54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081449.2 | NP_001074918.2 | 3 | 1102 |
Gene Symbol: Vmn2r55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104645.1 | NP_001098115.1 | 3 | 1102 | ||
GenBank mRNA | AB919108.1 | - | 3 | 1102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
