siRNA Details
Gene Symbol: Mcrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164156.1 | NP_001157628.1 | 7 | 863 | ||
NM_016766.3 | NP_058046.2 | 8 | 983 | |||
GenBank mRNA | AF015309.1 | - | 8 | 933 | ||
AK088116.1 | - | 8 | 948 | |||
AK158347.1 | - | 8 | 963 | |||
AK158611.1 | - | 4 | 590 | |||
AK167242.1 | - | 7 | 823 | |||
AK181698.1 | - | 3 | 281 | |||
AK184558.1 | - | 3 | 281 | |||
AK184749.1 | - | 3 | 281 | |||
BC003746.1 | - | 8 | 967 | 3486681 | ||
BC085099.1 | - | 8 | 951 | 6391692 | ||
BC108340.1 | - | 8 | 958 | 4504579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
