siRNA Details
Gene Symbol: Obox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145707.3 | NP_663753.3 | 5 | 1548 | ||
GenBank mRNA | AK139750.1 | - | 5 | 1549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
