siRNA Details
Gene Symbol: Gm13139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083918.1 | NP_001077387.1 | 7 | 2724 | ||
GenBank mRNA | BC024323.1 | - | 7 | 2724 | 4242170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
