siRNA Details
Gene Symbol: Snrpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026506.2 | NP_080782.1 | 4 | 342 | ||
GenBank mRNA | AK011129.1 | - | 4 | 341 | ||
AK012802.1 | - | 4 | 331 | |||
AK013235.1 | - | 4 | 343 | |||
AK017852.1 | - | 4 | 691 | |||
BC027499.1 | - | 4 | 315 | 1430048 | ||
BC051470.1 | - | 4 | 387 | 1244608 | ||
BC094411.1 | - | 4 | 329 | 1380524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
