siRNA Details
Gene Symbol: Vmn1r135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166747.1 | NP_001160219.1 | 1 | 791 |
Gene Symbol: Vmn1r128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166739.1 | NP_001160211.1 | 1 | 791 |
Gene Symbol: Vmn1r163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166755.1 | NP_001160227.1 | 1 | 791 |
