siRNA Details
Gene Symbol: Igkv10-96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC111442.1 | - | 60 | |||
BC111582.1 | - | 60 | ||||
BC128281.1 | - | 1 | 175 | 40099836 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
