siRNA Details
Gene Symbol: 4930453N24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026273.2 | NP_080549.1 | 3 | 969 | ||
GenBank mRNA | AK002578.1 | - | 3 | 964 | ||
AK075826.1 | - | 3 | 953 | |||
AK161311.1 | - | 3 | 970 | |||
BC020029.1 | - | 3 | 958 | 4012943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
