siRNA Details
Gene Symbol: Zfp534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127188.2 | NP_001120660.1 | 4 | 751 |
Gene Symbol: Gm13051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037926.2 | NP_001033015.1 | 4 | 706 | ||
GenBank mRNA | BC108336.1 | - | 4 | 684 | 30462068 |
