siRNA Details
Gene Symbol: LOC100038947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173459.2 | NP_001166930.1 | 6 | 1302 | ||
GenBank mRNA | AK029805.1 | - | 3 | 373 | ||
BC039636.1 | - | 1131 | ||||
BY551146.1 | - | 3 | 286 |
Gene Symbol: Sirpb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB112024.1 | - | 1287 | |||
AB112025.1 | - | 4 | 736 | |||
AK036935.1 | - | 1263 |
Gene Symbol: Sirpb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173460.1 | NP_001166931.1 | 6 | 1343 | ||
GenBank mRNA | AK154879.1 | - | 6 | 1344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
