siRNA Details
Gene Symbol: Syce1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048145.1 | NP_001041610.1 | 4, 5 | 293 | ||
NM_001286494.1 | NP_001273423.1 | 3, 4 | 230 | |||
NM_029139.1 | NP_083415.1 | 4, 5 | 293 | |||
GenBank mRNA | AK015606.1 | - | 4, 5 | 289 | ||
AY899935.1 | - | 4, 5 | 293 | |||
AY899936.1 | - | 4, 5 | 293 | |||
BB616021.1 | - | 4, 5 | 273 | |||
BC100478.1 | - | 3, 4 | 211 | |||
BC141326.1 | - | 4, 5 | 293 | 9055938 | ||
BC141347.1 | - | 4, 5 | 293 | 9055959 |
