siRNA Details
Gene Symbol: Lipo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013770.3 | NP_001013792.2 | 11 | 2150 | ||
GenBank mRNA | AK135585.1 | - | 5 | 1301 | ||
AK139790.1 | - | 11 | 2151 | |||
AK170332.1 | - | 11 | 2084 | |||
BC052506.1 | - | 4 | 905 | |||
BC145463.1 | - | 4 | 866 | |||
EU234002.1 | - | 4 | 2730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
