siRNA Details
Gene Symbol: Mettl7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081471.1 | NP_001074940.1 | 2 | 609 | ||
GenBank mRNA | AB253945.1 | - | 2 | 609 |
Gene Symbol: Mettl7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199477.2 | NP_955771.2 | 2 | 618 | ||
GenBank mRNA | AB099518.1 | - | 2 | 655 | ||
AB253944.1 | - | 2 | 609 | |||
AK143923.1 | - | 2 | 619 | |||
BC087869.1 | - | 2 | 618 | 30304748 | ||
BC090971.1 | - | 2 | 618 | 30314995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
