siRNA Details
Gene Symbol: Gm9112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177365.1 | NP_001170836.1 | 2 | 152 | ||
GenBank mRNA | CJ042093.1 | - | 2 | 153 | ||
CJ042401.1 | - | 2 | 118 |
Gene Symbol: 1700011M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005877.1 | - | 2 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
