siRNA Details
Gene Symbol: Acp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110239.1 | NP_001103709.1 | 3 | 302 | ||
GenBank mRNA | AK014603.1 | - | 3 | 273 | ||
AK018329.1 | - | 3 | 281 | |||
AK019186.1 | - | 3 | 249 | |||
AK136144.1 | - | 3 | 279 | |||
AK152429.1 | - | 3 | 280 | |||
AK170284.1 | - | 3 | 258 | |||
AK217793.1 | - | 1 | 70 | |||
BC039744.1 | - | 3 | 303 | 3156157 | ||
BC093527.1 | - | 3 | 229 | 5137663 | ||
BC112413.1 | - | 3 | 241 | 40060984 | ||
BC145199.1 | - | 3 | 241 | 9053750 | ||
Y17343.1 | - | 3 | 222 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | Y17344.1 | - | 483 | Chr. - - [-] on Build |

More Information
Other Species Targeted

Gene Ontology Categories:
