siRNA Details
Gene Symbol: Fbxw28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177419.1 | NP_001170890.1 | 10 | 1499 | ||
GenBank mRNA | AK143322.1 | - | 10 | 1495 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxw28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177419.1 | NP_001170890.1 | 10 | 1499 | ||
GenBank mRNA | AK143322.1 | - | 10 | 1495 |