siRNA Details
Gene Symbol: Gm21637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270685.1 | NP_001257614.1 | 1 | 490 | ||
GenBank mRNA | BC117726.1 | - | 1 | 490 | 40089629 |
Gene Symbol: Spin2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029106.2 | NP_083382.2 | 1 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
