siRNA Details
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204156.1 | NP_001191085.1 | 1 | 340 | ||
GenBank mRNA | AF002670.1 | - | 1 | 136 | ||
AK031555.1 | - | 1 | 780 | |||
BC047929.1 | - | 1 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
