siRNA Details
Gene Symbol: Gstm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008183.3 | NP_032209.1 | 7 | 537 | ||
GenBank mRNA | AK002845.1 | - | 7 | 554 | ||
BC019426.1 | - | 7 | 506 | |||
BC037068.1 | - | 7 | 561 | 4990201 | ||
J04696.1 | - | 7 | 498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
