siRNA Details
Gene Symbol: Zfp850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254951.1 | NP_001241880.1 | 6 | 6470 | ||
GenBank mRNA | AK081702.1 | - | 1 | 3803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
