siRNA Details
Gene Symbol: Gm14625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220498.1 | NP_001207427.1 | 3 | 255 | ||
GenBank mRNA | BC145074.1 | - | 255 | 9053622 | ||
BC171942.1 | - | 3 | 255 | 9054228 |
Gene Symbol: Gm16404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220497.1 | NP_001207426.1 | 3 | 255 | ||
GenBank mRNA | BC171941.1 | - | 3 | 255 | 9054227 |
Gene Symbol: Gm16405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166646.1 | NP_001160118.1 | 257 |
Gene Symbol: Gm16430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166601.1 | NP_001160073.1 | 3 | 257 | ||
GenBank mRNA | BC108390.1 | - | 3 | 257 |
Gene Symbol: Slxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029181.1 | NP_083457.1 | 3 | 292 | ||
GenBank mRNA | AK015913.1 | - | 3 | 293 | ||
BC108389.1 | - | 260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
