siRNA Details
Gene Symbol: SECTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003004.2 | NP_002995.1 | 5 | 1966 | ||
XM_005256392.3 | XP_005256449.1 | 5 | 1750 | |||
XM_011523588.2 | XP_011521890.1 | 5 | 1997 | |||
GenBank mRNA | AK223068.1 | - | 5 | 1659 | ||
AK223104.1 | - | 5 | 1690 | |||
BC017716.1 | - | 5 | 1660 | 4413387 | ||
U77643.1 | - | 5 | 1685 |
