siRNA Details
Gene Symbol: TRAPPC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002166.1 | 2 | 556 | |||
GenBank mRNA | AF058918.1 | - | 2 | 556 | ||
BU556832.1 | - | 2 | 550 | |||
CR456859.1 | - | 1 | 344 |
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011658.3 | NP_001011658.1 | 6 | 616 | ||
NM_001128835.2 | NP_001122307.2 | 6 | 610 | |||
NM_014563.5 | NP_055378.1 | 5 | 474 | |||
XM_011545565.1 | XP_011543867.1 | 7 | 684 | |||
XM_011545566.2 | XP_011543868.1 | 6 | 540 | |||
GenBank mRNA | AK313736.1 | - | 6 | 556 | ||
BC016915.1 | - | 5 | 437 | 4432196 | ||
BC052618.1 | - | 5 | 437 | 6581382 | ||
CB962763.1 | - | 5 | 459 | |||
DA344866.1 | - | 5 | 474 | |||
DA542477.1 | - | 6 | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
