siRNA Details
Gene Symbol: TRAPPC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002166.1 | 2 | 511 | |||
GenBank mRNA | AF058918.1 | - | 2 | 511 | ||
BU556832.1 | - | 2 | 505 | |||
CR456859.1 | - | 1 | 299 |
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011658.3 | NP_001011658.1 | 5 | 571 | ||
NM_001128835.2 | NP_001122307.2 | 5 | 565 | |||
NM_014563.5 | NP_055378.1 | 4 | 429 | |||
XM_011545565.1 | XP_011543867.1 | 6 | 639 | |||
XM_011545566.2 | XP_011543868.1 | 5 | 495 | |||
GenBank mRNA | AK098253.1 | - | 3 | 2578 | ||
AK310542.1 | - | 4 | 919 | |||
AK313736.1 | - | 5 | 511 | |||
BC016915.1 | - | 4 | 392 | 4432196 | ||
BC052618.1 | - | 4 | 392 | 6581382 | ||
BI821518.1 | - | 4 | 403 | |||
CB962763.1 | - | 4 | 414 | |||
CN430081.1 | - | 1 | 50 | |||
DA344866.1 | - | 4 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
