siRNA Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301781.1 | NP_001288710.1 | 11 | 2646 | ||
NM_207232.2 | NP_997115.1 | 10 | 2463 | |||
GenBank mRNA | AB185283.1 | - | 3 | 247 | ||
AK030415.1 | - | 10 | 2456 | |||
AK038922.1 | - | 10 | 2376 | |||
AK132781.1 | - | 11 | 2555 | |||
BC066081.1 | - | 10 | 2364 | 30355705 | ||
BC068267.1 | - | 3 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
