siRNA Details
Gene Symbol: Llph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025431.2 | NP_079707.1 | 3 | 694 | ||
GenBank mRNA | AK004503.1 | - | 3 | 694 | ||
AK007375.1 | - | 3 | 677 | |||
AK158700.1 | - | 3 | 672 | |||
BC024098.1 | - | 3 | 695 | 5148362 | ||
BC096681.1 | - | 3 | 693 | 1332438 | ||
BC106103.1 | - | 677 | 3661552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
