siRNA Details
Gene Symbol: Psma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011967.3 | NP_036097.1 | 7, 8 | 906 | ||
GenBank mRNA | AF019661.1 | - | 7, 8 | 621 | ||
AK143468.1 | - | 7, 8 | 908 | |||
AK160530.1 | - | 7, 8 | 666 | |||
AK160550.1 | - | 7, 8 | 660 | |||
AK160741.1 | - | 7, 8 | 669 | |||
AK161680.1 | - | 7, 8 | 654 | |||
BC007181.1 | - | 7, 8 | 653 | |||
BC010709.1 | - | 7, 8 | 642 | 2648635 | ||
BC083342.1 | - | 7, 8 | 639 | 5700963 | ||
BC108368.1 | - | 7, 8 | 636 | 3596530 |
