siRNA Details
Gene Symbol: Zfp146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011980.3 | NP_036110.1 | 2 | 1277 | ||
GenBank mRNA | AJ224763.1 | - | 2 | 1240 | ||
AK047696.1 | - | 2 | 1285 | |||
BC127038.1 | - | 1 | 718 | 40051938 | ||
BC127039.1 | - | 1 | 719 | 40051941 |
