siRNA Details
Gene Symbol: Esp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244651.1 | NP_001231580.1 | 3 | 204 | ||
GenBank mRNA | AB306993.1 | - | 3 | 204 |
Gene Symbol: Esp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255977.1 | NP_001242906.1 | 3 | 286 | ||
GenBank mRNA | AB306995.1 | - | 3 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
