siRNA Details
Gene Symbol: Tigar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177003.5 | NP_795977.1 | 6 | 596 | ||
GenBank mRNA | AK036082.1 | - | 6 | 587 | ||
AK078900.1 | - | 6 | 588 | |||
AK145896.1 | - | 6 | 581 | |||
AK163290.1 | - | 6 | 572 | |||
BC147705.1 | - | 6 | 586 | 9087464 | ||
BC147718.1 | - | 6 | 586 | 9087477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
