siRNA Details
Gene Symbol: Mfap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026220.4 | NP_080496.1 | 2 | 437 | ||
GenBank mRNA | AK012688.1 | - | 2 | 423 | ||
AK014483.1 | - | 2 | 430 | |||
AK032586.1 | - | 2 | 438 | |||
AK050561.1 | - | 2 | 438 | |||
AK077604.1 | - | 2 | 431 | |||
AK088766.1 | - | 2 | 434 | |||
AK161000.1 | - | 2 | 438 | |||
AK168801.1 | - | 2 | 443 |
Gene Symbol: Mfap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081975.3 | NP_001075444.1 | 2 | 433 | ||
GenBank mRNA | AK011317.1 | - | 2 | 431 | ||
AK018514.1 | - | 2 | 434 | |||
AK088946.1 | - | 440 | ||||
AK169029.1 | - | 441 | ||||
BC005728.1 | - | 2 | 394 | 3595848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
