siRNA Details
Gene Symbol: Tmsb15l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207267.4 | NP_997150.1 | 3 | 269 | ||
GenBank mRNA | AK029707.1 | - | 3 | 246 | ||
BB623661.1 | - | 3 | 265 | |||
BC096054.1 | - | 3 | 202 | 3373135 | ||
BE625563.1 | - | 3 | 194 |
Gene Symbol: Tmsb15b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080967.4 | NP_001074436.1 | 2 | 129 | ||
GenBank mRNA | CN836907.1 | - | 115 | |||
CN842750.1 | - | 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
