siRNA Details
Gene Symbol: LOC100911545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749828.2 | XP_003749876.1 | 2406 |
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012488.2 | NP_036620.2 | 17 | 2100 | ||
GenBank mRNA | AY919611.1 | - | 3 | 232 | ||
BC098922.1 | - | 17 | 2100 | 7458629 | ||
FQ223246.1 | - | 17 | 2122 | |||
FQ228339.1 | - | 17 | 2299 | |||
J02635.1 | - | 17 | 2122 |
