siRNA Details
Gene Symbol: Acly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111095.1 | NP_001104565.1 | 24 | 2923 | ||
NM_016987.2 | NP_058683.2 | 25 | 2953 | |||
XM_006247254.2 | XP_006247316.1 | 25 | 2946 | |||
GenBank mRNA | BC100618.1 | - | 24 | 2915 | 7934668 | |
J05210.1 | - | 25 | 2901 |
