siRNA Details
Gene Symbol: Add2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109880.1 | NP_001103350.1 | 12, 13 | 1694 | ||
NM_012491.2 | NP_036623.1 | 12, 13 | 1694 | |||
GenBank mRNA | M63894.1 | - | 12, 13 | 1694 |

More Information
Other Species Targeted

Gene Ontology Categories:
