siRNA Details
Gene Symbol: Adh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF508795.1 | - | 5 | 535 | ||
BC062403.1 | - | 5 | 507 | 5621394 | ||
CK480694.1 | - | 5 | 500 | |||
FQ209590.1 | - | 5 | 518 | |||
FQ218571.1 | - | 5 | 520 | |||
FQ220674.1 | - | 5 | 518 | |||
M15327.1 | - | 5 | 491 | |||
X72792.1 | - | 3 | 321 |
