siRNA Details
Gene Symbol: Agtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012494.3 | NP_036626.1 | 1 | 150 | ||
XM_006257432.2 | XP_006257494.1 | 3 | 492 | |||
XM_008773399.1 | XP_008771621.1 | 3 | 301 | |||
GenBank mRNA | BC161802.1 | - | 2 | 395 | 9102491 | |
D16840.1 | - | 3 | 289 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
