siRNA Details
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012496.2 | NP_036628.2 | 7 | 710 | ||
GenBank mRNA | BC081697.1 | - | 7 | 710 | 7109764 | |
FQ209505.1 | - | 7 | 730 | |||
FQ209538.1 | - | 7 | 730 | |||
FQ209583.1 | - | 7 | 730 | |||
FQ209594.1 | - | 7 | 730 | |||
FQ209621.1 | - | 7 | 728 | |||
FQ209639.1 | - | 7 | 730 | |||
FQ209643.1 | - | 7 | 727 | |||
FQ209782.1 | - | 7 | 730 | |||
FQ209819.1 | - | 7 | 740 | |||
FQ210002.1 | - | 7 | 730 | |||
FQ210453.1 | - | 5 | 437 | |||
FQ210849.1 | - | 7 | 730 | |||
FQ211083.1 | - | 7 | 730 | |||
FQ211159.1 | - | 7 | 730 | |||
FQ211293.1 | - | 5 | 408 | |||
FQ218139.1 | - | 7 | 730 | |||
FQ218217.1 | - | 7 | 730 | |||
FQ218348.1 | - | 7 | 736 | |||
FQ218402.1 | - | 7 | 732 | |||
FQ218551.1 | - | 7 | 730 | |||
FQ218592.1 | - | 3 | 228 | |||
FQ218665.1 | - | 7 | 730 | |||
FQ218885.1 | - | 7 | 729 | |||
FQ218911.1 | - | 7 | 730 | |||
FQ219077.1 | - | 5 | 414 | |||
FQ219116.1 | - | 7 | 730 | |||
FQ219236.1 | - | 7 | 729 | |||
FQ219261.1 | - | 7 | 730 | |||
FQ219453.1 | - | 7 | 730 | |||
FQ219555.1 | - | 7 | 730 | |||
FQ219574.1 | - | 7 | 730 | |||
FQ219640.1 | - | 7 | 728 | |||
FQ219646.1 | - | 7 | 730 | |||
FQ219703.1 | - | 7 | 729 | |||
M10149.1 | - | 7 | 721 | |||
V01223.1 | - | 2 | 101 |

More Information
Other Species Targeted

Gene Ontology Categories:
